missing translation for 'onlineSavingsMsg'
Learn More

Allylchlorodimethylsilane, 94%

Product Code. 11425886
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425886 25 g 25g
11415886 5 g 5g
11374609 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425886 Supplier Thermo Scientific Alfa Aesar Supplier No. B22237.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4028-23-3 | C5H11ClSi | 134.678 g/mol

Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1?,4,4?-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1′,4,4′-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

Solubility
Soluble in most organic solvents; reacts with alcohols, amines, and water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4028-23-3
Molecular Formula C5H11ClSi
Molecular Weight (g/mol) 134.678
MDL Number MFCD00000506
InChI Key KMVZWUQHMJAWSY-UHFFFAOYSA-N
Synonym allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7
PubChem CID 77646
IUPAC Name chloro-dimethyl-prop-2-enylsilane
SMILES C[Si](C)(CC=C)Cl

Specifications

Boiling Point 110°C to 112°C
Quantity 25 g
Flash Point 5°C (41°F)
Refractive Index 1.429
Sensitivity Moisture sensitive
Density 0.896
UN Number UN2985
Beilstein 1737978
Solubility Information Soluble in most organic solvents; reacts with alcohols,amines,and water.
Formula Weight 134.68
Percent Purity 97%
Chemical Name or Material Allylchlorodimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.