Learn More
Allylbenzene, 98%
Description
Allylbenzene is used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Allylbenzene is used in organic synthesis.
Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼-50°C |
| Density | 0.892 |
| Boiling Point | 158°C to 160°C |
| Flash Point | 33°C (91°F) |
| Odor | Aromatic |
| Refractive Index | 1.51 |
| Quantity | 25 g |
| UN Number | UN3295 |
| Beilstein | 1098501 |
| Solubility Information | Insoluble in water. Soluble in alcohol,ether and benzene. |
| Show More |
Safety and Handling
- Allylbenzene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- MIXTURE LIST-Contain: Allylbenzene
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.