missing translation for 'onlineSavingsMsg'
Learn More

Allylamine, 98%

Product Code. 10338310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338310 250 mL Glass bottle 250mL
10637202 50 mL Glass bottle 50mL
10013473 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10338310 Supplier Thermo Scientific Acros Supplier No. 220732500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 107-11-9 | C3H7N | 57.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-11-9
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.10
MDL Number MFCD00008199
InChI Key VVJKKWFAADXIJK-UHFFFAOYSA-N
Synonym allylamine, 2-propen-1-amine, monoallylamine, 3-aminopropylene, 3-aminopropene, allyl amine, 2-propenamine, 3-amino-1-propene, 2-propenylamine, polyallylamine
PubChem CID 7853
IUPAC Name prop-2-en-1-amine
SMILES NCC=C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -88°C
Color White
Density 0.7630g/mL
Boiling Point 53°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2NH2
Refractive Index 1.4205 to 1.4225
Quantity 250 mL
Beilstein 04,205
Merck Index 15,278
Specific Gravity 0.763
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether
Viscosity 0.44 mPa.s (25°C)
Formula Weight 57.09
Percent Purity 98%
Grade Extra Pure
Physical Form Crystalline or Granular Powder
Chemical Name or Material Allylamine, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.