missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Product Code. 11321707
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321707 10 g 10g
11331707 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321707 Supplier Thermo Scientific Alfa Aesar Supplier No. L04771.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5296-64-0
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00014957
InChI Key QGNRLAFFKKBSIM-UHFFFAOYSA-N
Synonym allyl phenyl sulfide, allylthio benzene, allyl phenyl sulfane, benzene, 2-propenylthio, allylphenyl sulfide, phenyl allyl sulfide, allylphenylthioether, allyl phenyl ;sulfane, sulfide, allyl phenyl
PubChem CID 79180
IUPAC Name prop-2-enylsulfanylbenzene
SMILES C=CCSC1=CC=CC=C1

Specifications

Density 1.024
Boiling Point 223°C to 225°C
Flash Point >98°C (208°F)
Odor Unpleasant
Refractive Index 1.574
Quantity 10 g
Beilstein 1856940
Sensitivity Air Sensitive
Formula Weight 150.24
Percent Purity 97%
Chemical Name or Material Allyl phenyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.