missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Phenyl Ether, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Allyl Phenyl Ether |
| CAS | 1746-13-0 |
| Molecular Formula | C9H10O |
| Quantity | 1 g |
| Synonym | Allyloxybenzene, NSC 4746, Phenyl 2-Propenyl Ether, Phenyl Allyl Ether, Phenylpropenyl Ether, (2-Propenyloxy)benzene, 1-(2-Propenyloxy)benzene, 1-Phenoxy-2-propene, 3-Phenoxy-1-propene, 3-Phenoxypropene, Ether Allyl Phenyl, (2-Propenyloxy)benzene |
| InChI Formula | InChI=1S/C9H10O/c1-2-8-10-9-6-4-3-5-7-9/h2-7H,1,8H2 |
| SMILES | C=CCOc1ccccc1 |
| IUPAC Name | prop-2-enoxybenzene |
| Molecular Weight (g/mol) | 134.18 |
| Formula Weight | 134.0732 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction