missing translation for 'onlineSavingsMsg'
Learn More

Allyl methyl ether, 94%, Thermo Scientific Chemicals

Catalog Number 11313457
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
627-40-7
C4H8O
72.11
MFCD00008649
FASUFOTUSHAIHG-UHFFFAOYSA-N
allyl methyl ether, 1-propene, 3-methoxy, 3-methoxy-1-propene, ether, allyl methyl, 3-methoxypropene, methyl allyl ether, allylmethylether, 4-oxapent-1-ene, 1-methoxy-2-propene, methylallyl ether
69392
3-methoxyprop-1-ene
COCC=C
This item is not returnable. View return policy
627-40-7
C4H8O
72.11
MFCD00008649
FASUFOTUSHAIHG-UHFFFAOYSA-N
allyl methyl ether, 1-propene, 3-methoxy, 3-methoxy-1-propene, ether, allyl methyl, 3-methoxypropene, methyl allyl ether, allylmethylether, 4-oxapent-1-ene, 1-methoxy-2-propene, methylallyl ether
69392
3-methoxyprop-1-ene
COCC=C

Allyl methyl ether it used as pharmaceutical intermediate. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.77 g/mL
Boiling Point 42°C to 43°C
Flash Point -23°C (-94°F)
Odor Ether-like
Refractive Index 1.3803
Quantity 10 g
UN Number UN3271
Formula Weight 72.1
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Allyl methyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'