missing translation for 'onlineSavingsMsg'
Learn More

Allyl methacrylate, 98%, stabilized

Product Code. 10328640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10328640 500 mL Glass bottle 500mL
11914961 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10328640 Supplier Thermo Scientific Acros Supplier No. 215875000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-05-9 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-05-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00008592
InChI Key FBCQUCJYYPMKRO-UHFFFAOYSA-N
Synonym allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech
PubChem CID 7274
IUPAC Name prop-2-enyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC=C

Specifications

Acidity 0.05% max.
Melting Point -65.0°C
Density 0.9300g/mL
Boiling Point 144.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH=CH2
Refractive Index 1.4350 to 1.4370
Quantity 500 mL
Beilstein 02,III,129
Specific Gravity 0.93
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: miscible with almost all organic solvents
Stabilizer 200 to 250ppm MEHQ
Viscosity 1 mPa.s (20°C)
Formula Weight 126.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl methacrylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.