missing translation for 'onlineSavingsMsg'
Learn More

Allyl isocyanate, 96%

Product Code. 15479079
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479079 1 g 1g
15489079 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479079 Supplier Thermo Scientific Alfa Aesar Supplier No. L18710.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1476-23-9 | C4H5NO | 83.09 g/mol

It finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-23-9
Molecular Formula C4H5NO
Molecular Weight (g/mol) 83.09
MDL Number MFCD00002044
InChI Key HXBPYFMVGFDZFT-UHFFFAOYSA-N
PubChem CID 15123
IUPAC Name 3-isocyanatoprop-1-ene
SMILES C=CCN=C=O

Specifications

Density 0.94
Boiling Point 87°C to 89°C
Flash Point 6°C (43°F)
Odor Pungent
Refractive Index 1.4185
Quantity 1 g
UN Number UN2478
Beilstein 506106
Sensitivity Moisture sensitive; May form a precipitate
Synonym allyl isocyanate, 1-propene, 3-isocyanato, 3-isocyanato-1-propene, propene, 3-isocyanato, allylisocyanate, isocyanic acid, allyl ester, 1-propene,3-isocyanato, prop-2-enisocyanate, 2-propenyl isocyanate, isocyanicacid,allylester
Solubility Information Hydrolyzes in water.
Formula Weight 83.09
Percent Purity 98%
Chemical Name or Material Allyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Allyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.