missing translation for 'onlineSavingsMsg'
Learn More

Allyl heptanoate, 97%, Thermo Scientific™

Product Code. p-7045426
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11312936

Brand: Thermo Scientific Alfa Aesar L10671.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-19-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00038341
InChI Key SJWKGDGUQTWDRV-UHFFFAOYSA-N
Synonym allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester
PubChem CID 8878
IUPAC Name prop-2-enyl heptanoate
SMILES CCCCCCC(=O)OCC=C

Specifications

Density 0.882
Boiling Point 210°C to 212°C
Flash Point 74°C (165°F)
Linear Formula CH3(CH2)5CO2CH2CH=CH2
Refractive Index 1.4291
Quantity 500 g
UN Number UN2810
Beilstein 8544440
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material Allyl heptanoate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.