missing translation for 'onlineSavingsMsg'
Learn More

Allyl glycidyl ether, 97%

Product Code. 11382537
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382537 500 g 500g
11372537 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382537 Supplier Thermo Scientific Alfa Aesar Supplier No. L11634.36
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-92-3 | C6H10O2 | 114.144 g/mol

Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

Solubility
Soluble in water (140 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

Melting Point -100°C
Density 0.967
Boiling Point 50°C to 51°C (15 mmHg)
Flash Point 47°C (134°F)
Odor Ether-like
Refractive Index 1.435
Quantity 500 g
UN Number UN2219
Beilstein 105871
Solubility Information Soluble in water (140g/L at 20°C).
Formula Weight 114.15
Percent Purity 97%
Chemical Name or Material Allyl glycidyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.