missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloroformate, 97%

Product Code. 10472221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10472221 50 mL Glass bottle 50mL
10781335 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10472221 Supplier Thermo Scientific Acros Supplier No. 221740500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 2937-50-0 | C4H5ClO2 | 120.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2937-50-0
Molecular Formula C4H5ClO2
Molecular Weight (g/mol) 120.54
MDL Number MFCD00000648
InChI Key CAEWJEXPFKNBQL-UHFFFAOYSA-N
Synonym allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech
PubChem CID 18052
IUPAC Name prop-2-enyl carbonochloridate
SMILES C=CCOC(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown-Red to Yellow
Density 1.1300g/mL
Boiling Point 109.0°C to 110.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2CH2CH=CH2
Refractive Index 1.4213 to 1.4233
Quantity 50 mL
Beilstein 03, 12
Fieser 12,15; 13,9
Specific Gravity 1.13
Solubility Information Solubility in water: decomposes
Formula Weight 120.54
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.