missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloroacetate, 98%

Product Code. 11302477
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302477 5 g 5g
11312477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302477 Supplier Thermo Scientific Alfa Aesar Supplier No. L11531.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2916-14-5 | C5H7ClO2 | 134.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2916-14-5
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.559
MDL Number MFCD00058936
InChI Key VMBJJCDVORDOCF-UHFFFAOYSA-N
Synonym allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester
PubChem CID 76206
IUPAC Name prop-2-enyl 2-chloroacetate
SMILES C=CCOC(=O)CCl

Specifications

Density 1.16
Boiling Point 162°C to 164°C
Flash Point 61°C (141°F)
Refractive Index 1.445
Quantity 5 g
UN Number UN2810
Beilstein 1747000
Formula Weight 134.56
Percent Purity 98%
Chemical Name or Material Allyl chloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.