missing translation for 'onlineSavingsMsg'
Learn More

Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide

Product Code. 11419333
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419333 1000 g 1000g
11429333 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419333 Supplier Thermo Scientific Alfa Aesar Supplier No. A11766.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-95-6 | C3H5Br | 120.977 g/mol

Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

Solubility
Slightly miscible with water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkali metals, alkaline earth metals, amides, amines and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-95-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00000244
InChI Key BHELZAPQIKSEDF-UHFFFAOYSA-N
Synonym allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
PubChem CID 7841
IUPAC Name 3-bromoprop-1-ene
SMILES C=CCBr

Specifications

Melting Point -119°C
Density 1.398
Boiling Point 70°C to 71°C
Flash Point −2°C (28°F)
Assay Percent Range 99%
Odor Pungent
Linear Formula CH2=CHCH2Br
Refractive Index 1.47
Quantity 1000 g
UN Number UN1099
Sensitivity Light sensitive
Merck Index 14,288
Solubility Information Slightly miscible with water.
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl bromide, Stabilized with 300 to 1000ppm Propylene oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl bromide, stabilized with 300-1000ppm Propylene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Allyl bromide, Propylene oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.