missing translation for 'onlineSavingsMsg'
Learn More

Allyl Alcohol, 99%, extra pure

Product Code. 10564004 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10564004 250 mL Glass bottle 250mL
10283980 1 L Glass bottle 1 L
11904621 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10564004 Supplier Thermo Scientific Acros Supplier No. 102862500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 107-18-6 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-18-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00002920
InChI Key XXROGKLTLUQVRX-UHFFFAOYSA-N
Synonym allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench
PubChem CID 7858
ChEBI CHEBI:16605
IUPAC Name prop-2-en-1-ol
SMILES OCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -129.0°C
Density 0.8500g/mL
Boiling Point 96.0°C to 98.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2OH
Refractive Index 1.4119 to 1.4139
Quantity 250 mL
Beilstein 01,436
Merck Index 15,277
Specific Gravity 0.85
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, chloroform and ether
Formula Weight 58.08
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Allyl alcohol, Extra Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl alcohol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.