missing translation for 'onlineSavingsMsg'
Learn More

Allyl alcohol, 98+%

Product Code. 11401960
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1000mL
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401960 100 mL 100mL
11670212 500 mL 500mL
15413107 1 L 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11401960 Supplier Thermo Scientific Alfa Aesar Supplier No. A15026.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 107-18-6 | C3H6O | 58.08 g/mol

Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, oleum and alkali metals. Polymerizes during prolonged storage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-18-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00002920
InChI Key XXROGKLTLUQVRX-UHFFFAOYSA-N
Synonym allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench
PubChem CID 7858
ChEBI CHEBI:16605
IUPAC Name prop-2-en-1-ol
SMILES OCC=C

Specifications

Melting Point -129°C
Density 0.852
Boiling Point 95°C to 97°C
Flash Point 22°C (71°F)
Assay Percent Range ≥98%
Odor Pungent
Linear Formula CH2=CHCH2OH
Refractive Index 1.413
Quantity 100 mL
UN Number UN1098
Beilstein 605307
Merck Index 14,286
Solubility Information Miscible with water.
Formula Weight 58.08
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Allyl alcohol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl alcohol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Allyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.