missing translation for 'onlineSavingsMsg'
Learn More

Allyl acrylate, 95%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

Catalog Number 15421997
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
999-55-3
C6H8O2
112.128
MFCD00014949
QTECDUFMBMSHKR-UHFFFAOYSA-N
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
13835
prop-2-enyl prop-2-enoate
C=CCOC(=O)C=C
This item is not returnable. View return policy
999-55-3
C6H8O2
112.128
MFCD00014949
QTECDUFMBMSHKR-UHFFFAOYSA-N
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
13835
prop-2-enyl prop-2-enoate
C=CCOC(=O)C=C

Allyl acrylate is a monofunctional monomer which function as a chain stopper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.935 g/mL
Boiling Point 46°C to 47°C (40 mmHg)
Flash Point 32°C (89°F)
Odor Pungent
Refractive Index 1.436
Quantity 25 g
UN Number UN2929
Beilstein 1743576
Solubility Information Slightly soluble in water.
Formula Weight 112.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Allyl acrylate, Stabilized with 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'