missing translation for 'onlineSavingsMsg'
Learn More

Allyl Acetate, TRC

Product Code. 30309099
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30309099

Brand: Toronto Research Chemicals TRCA54901550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allyl Acetate
CAS 591-87-7
Molecular Formula C5H8O2
Quantity 50 g
Synonym Acetic Acid 2-Propen-1-yl Ester, Acetic Acid 2-Propenyl Ester, Acetic Acid Allyl Ester, Allyl Alcohol Acetate, 2-Propen-1-ol Acetate, 3-Acetoxy-1-propene, 3-Acetoxypropene, NSC 7612
InChI Formula InChI=1S/C5H8O2/c1-3-4-7-5(2)6/h3H,1,4H2,2H3
SMILES CC(=O)OCC=C
IUPAC Name prop-2-enyl acetate
Molecular Weight (g/mol) 100.12
Formula Weight 100.0524
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.