missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Acetate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Allyl Acetate |
| CAS | 591-87-7 |
| Molecular Formula | C5H8O2 |
| Quantity | 5 g |
| Synonym | Acetic Acid 2-Propen-1-yl Ester, Acetic Acid 2-Propenyl Ester, Acetic Acid Allyl Ester, Allyl Alcohol Acetate, 2-Propen-1-ol Acetate, 3-Acetoxy-1-propene, 3-Acetoxypropene, NSC 7612 |
| InChI Formula | InChI=1S/C5H8O2/c1-3-4-7-5(2)6/h3H,1,4H2,2H3 |
| SMILES | CC(=O)OCC=C |
| IUPAC Name | prop-2-enyl acetate |
| Molecular Weight (g/mol) | 100.12 |
| Formula Weight | 100.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction