missing translation for 'onlineSavingsMsg'
Learn More

Allyl acetate, 98%

Product Code. 10499100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10499100 500 mL 500mL
10489100 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10499100 Supplier Thermo Scientific Acros Supplier No. 180655000
  • ONLINE EXCLUSIVE PRICE
    £89.00
    £73.65 / 500mL
    Save £15.35
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 591-87-7 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-87-7
MDL Number MFCD00008721
InChI Key FWZUNOYOVVKUNF-UHFFFAOYSA-N
Synonym allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i
PubChem CID 11584
IUPAC Name prop-2-enyl acetate
SMILES CC(=O)OCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -96.0°C
Color Colorless
Density 0.9200g/mL
Boiling Point 103.0°C to 104.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C5H8O2
Linear Formula CH3CO2CH2CH=CH2
Refractive Index 1.4030 to 1.4050
Quantity 500 mL
Beilstein 02, 136
Merck Index 15, 276
Specific Gravity 0.92
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 100.12
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl acetate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allyl acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.