missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl acetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
Description
Ally acetate is used in industrial intermediate. Used in synthesis of colophony and bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 6°C |
Density | 0.928 g/mL |
Boiling Point | 103°C to 104°C |
Refractive Index | 1.4042 |
Quantity | 250 mL |
UN Number | UN2333 |
Merck Index | 14,285 |
Solubility Information | Soluble in ethanol. Miscible with ethanol and ether. Slightly soluble in water. |
Formula Weight | 100.12 |
Percent Purity | 99% |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'