missing translation for 'onlineSavingsMsg'
Learn More

Grace™ Alltech™ Vydac™ 214TP C4 Column

A polymerically bonded endcapped n-butyl reversed-phase based on 300Å TP silica

Brand:  Grace™ 214TP5315

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 11359950

  • POA / Each
Estimated Shipment: 10-06-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Polymeric bonded phases have exceptionally long column lifetime and negligible phase leaching
  • Extensive applications library based on over two decade's experience

Specifications

Specifications

3.2 mm
150 mm
5 μm
60 to 110 m2/g
214 TP
3%
Yes
Glycoproteins, Hemoglobin Variants, Histones, Insulin Variants, Membrane Proteins
Spherical
300 Å
L26
Silica
C4
Vydac™ TP C4 Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers