missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, SUS, QAC, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, SUS, QAC, Ø38x32mm
Specifications
Specifications
| Chemical Name or Material | Silicon/Calcium/Niobium/Lead/Zinc/Zirconium/Boron/Aluminium/Nitrogen/Cobalt/Titanium/Antimony/Tantalum/Carbon/Arsenic/Chromium/Molybdenum/Phosphorus/Tin/Copper/Tungsten/Vanadium/Manganese/Nickel |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Silicon 0.1 %/Calcium (<0.0001) %/Niobium (<0.0001) %/Lead (<0.001) %/Zinc (<0.001) %/Zirconium (<0.001) %/Boron (0.0004) %/Aluminium (0.003) %/Nitrogen (0.007) %/Cobalt (0.01) %/Titanium (0.01) %/Antimony (0.02) %/Tantalum (0.02) %/Carbon (0.05) %/Arsenic (0.1) %/Chromium (0.1) %/Molybdenum (0.1) %/Phosphorus (0.1) %/Tin (0.1) %/Copper (0.6) %/Tungsten (0.7) %/Vanadium (1) %/Manganese (2) %/Nickel (5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction