missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Mo, AISI 4820, G48200, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Ni/Mo, AISI 4820, G48200, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Arsenic/Vanadium/Niobium/Titanium/Oxygen/Tungsten/Nitrogen/Phosphorus/Tin/Cobalt/Aluminium/Chromium/Carbon/Copper/Silicon/Molybdenum/Manganese/Nickel/Cadmium/Magnesium/Zinc/Calcium/Antimony/Tantalum/Lead/Boron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.0006 %/Vanadium 0.0015 %/Niobium 0.0016 %/Titanium 0.002 %/Oxygen 0.003 %/Tungsten 0.004 %/Nitrogen 0.005 %/Phosphorus 0.008 %/Tin 0.0084 %/Cobalt 0.012 %/Aluminium 0.0356 %/Chromium 0.144 %/Carbon 0.199 %/Copper 0.219 %/Silicon 0.223 %/Molybdenum 0.244 %/Manganese 0.617 %/Nickel 3.36 %/Cadmium (<0.001) %/Magnesium (<0.001) %/Zinc (<0.001) %/Calcium (<0.002) %/Antimony (<0.003) %/Tantalum (<0.005) %/Lead (<0.006) %/Boron (0.0004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction