missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Cr, AISI E52100, G52986, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Cr, AISI E52100, G52986, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Oxygen/Arsenic/Niobium/Nitrogen/Cobalt/Phosphorus/Titanium/Tin/Molybdenum/Aluminium/Nickel/Vanadium/Copper/Silicon/Manganese/Carbon/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.002 %/Arsenic 0.0029 %/Niobium 0.003 %/Nitrogen 0.003 %/Cobalt 0.006 %/Phosphorus 0.006 %/Titanium 0.006 %/Tin 0.0065 %/Molybdenum 0.017 %/Aluminium 0.024 %/Nickel 0.043 %/Vanadium 0.066 %/Copper 0.076 %/Silicon 0.248 %/Manganese 0.364 %/Carbon 1.03 %/Chromium 1.43 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction