missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, CLA7, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, CLA7, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Tin/Vanadium/Molybdenum/Niobium/Cobalt/Copper/Phosphorus/Zirconium/Chromium/Titanium/Nickel/Silicon/Aluminium/Carbon/Manganese/Cadmium/Lead/Zinc/Calcium/Magnesium/Selenium/Oxygen/Bismuth/Antimony/Tungsten/Arsenic/Nitrogen/Tantalum |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0004 %/Tin 0.002 %/Vanadium 0.002 %/Molybdenum 0.003 %/Niobium 0.004 %/Cobalt 0.005 %/Copper 0.005 %/Phosphorus 0.005 %/Zirconium 0.005 %/Chromium 0.009 %/Titanium 0.19 %/Nickel 0.197 %/Silicon 0.21 %/Aluminium 0.23 %/Carbon 0.4 %/Manganese 0.821 %/Cadmium (<0.0010) %/Lead (<0.0020) %/Zinc (<0.0020) %/Calcium (<0.0040) %/Magnesium (<0.0050) %/Selenium (<0.0050) %/Oxygen (<0.0060) %/Bismuth (<0.0100) %/Antimony (0.0007) %/Tungsten (0.002) %/Arsenic (0.005) %/Nitrogen (0.005) %/Tantalum (0.008) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction