missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Ø38x20mm
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Zinc/Zirconium/Phosphorus/Arsenic/Aluminium/Cobalt/Silicon/Vanadium/Copper/Molybdenum/Nickel/Chromium/Carbon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.003 %/Zinc 0.003 %/Zirconium 0.02 %/Phosphorus 0.025 %/Arsenic 0.03 %/Aluminium 0.04 %/Cobalt 0.05 %/Silicon 0.15 %/Vanadium 0.15 %/Copper 0.2 %/Molybdenum 0.3 %/Nickel 0.3 %/Chromium 0.35 %/Carbon 0.5 %/Manganese 1.6 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction