missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, 21⁄4Cr 1Mo, A182 Grade F22, K21590, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, 21⁄4Cr 1Mo, A182 Grade F22, K21590, 100g
Specifications
Specifications
| Chemical Name or Material | Bismuth/Cadmium/Selenium/Tantalum/Zinc/Boron/Calcium/Lead/Titanium/Magnesium/Antimony/Zirconium/Tungsten/Oxygen/Niobium/Arsenic/Vanadium/Cobalt/Nitrogen/Phosphorus/Tin/Aluminium/Nickel/Copper/Carbon/Silicon/Manganese/Molybdenum/Chromium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Bismuth (<0.001) %/Cadmium (<0.001) %/Selenium (<0.002) %/Tantalum (<0.002) %/Zinc (<0.002) %/Boron [(0.0003)] %/Calcium [(0.0003)] %/Lead [(0.0007)] %/Titanium [(0.0019)] %/Magnesium [(0.002)] %/Antimony [(0.003)] %/Zirconium [(0.003)] %/Tungsten [(0.0045)] %/Oxygen [0.0029] %/Niobium [0.004] %/Arsenic [0.005] %/Vanadium [0.0061] %/Cobalt [0.008] %/Nitrogen [0.008] %/Phosphorus [0.008] %/Tin [0.008] %/Aluminium [0.017] %/Nickel [0.085] %/Copper [0.143] %/Carbon [0.147] %/Silicon [0.255] %/Manganese [0.489] %/Molybdenum [0.96] %/Chromium [2.41] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction