missing translation for 'onlineSavingsMsg'
Learn More

Alloxan monohydrate, 98%

Product Code. 11394609
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
This item is not returnable. View return policy

Product Code. 11394609

Brand: Thermo Scientific Alfa Aesar A15324.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3237-50-1 | C4H4N2O5 | 160.085 g/mol

Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

Solubility
Soluble in water, ethanol, acetone, glacial acetic acid and methanol. Slightly soluble in chloroform, petroleum ether, toluene, ethyl acetate and acetic anhydride. Insoluble in ether.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3237-50-1
Molecular Formula C4H4N2O5
Molecular Weight (g/mol) 160.085
MDL Number MFCD00149399
InChI Key ZIIHZVKHFWOENY-UHFFFAOYSA-N
Synonym barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6
PubChem CID 312231
IUPAC Name 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
SMILES C1(=O)C(C(=O)NC(=O)N1)(O)O

Specifications

Melting Point ∼260°C (decomposition)
Beilstein 5309394
Merck Index 14,282
Formula Weight 160.09 (142.07 Anhydrous)
Percent Purity 98%
Quantity 100 g
Chemical Name or Material Alloxan monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.