missing translation for 'onlineSavingsMsg'
Learn More

Allopurinol, MedChemExpress

Product Code. 16480828
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480828 5 g 5g
16418428 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480828 Supplier Medchem Express Supplier No. HYB0219
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Allopurinol is a potent xanthine oxidase inhibitor (IC50 values of 0.2 to 50 μM). Allopurinol can be used for the research of hyperuricemia and gout. Antileishmanial effect.

Allopurinol is a potent xanthine oxidase inhibitor (IC50 values of 0.2 to 50 μM). Allopurinol can be used for the research of hyperuricemia and gout. Antileishmanial effect.

Specifications

Chemical Name or Material Allopurinol
Physical Form Solid
CAS 315-30-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C5H4N4O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 14 mg/mL (102.86 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (7.35 mM; ultrasonic and adjust pH to 11 with NaOH)
SMILES O=C1C2=C(NN=C2)N=CN1
Molecular Weight (g/mol) 136.11
Formula Weight 136.11
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.