missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allopregnanolone, ≥95%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| CAS | 516-54-1 |
| Color | White |
| Molecular Formula | C21H34O2 |
| MDL Number | MFCD00003677 |
| Quantity | 5 mg |
| Synonym | allopregnanolone, allopregnan-3alpha-ol-20-one, allotetrahydroprogesterone, 3alpha-hydroxy-5alpha-pregnan-20-one, brexanolone, 3a-hydroxy-5a-pregnan-20-one, unii-s39xz5qv8y, 3-alpha,5-alpha-pregnanolone, 3alpha-oh dhp, 5alpha-pregnan-3alpha-ol-20-one |
| InChI Key | AURFZBICLPNKBZ-SYBPFIFISA-N |
| SMILES | CC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | 1-[(3R,5S,8R,9S,10S,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone |
| Molecular Weight (g/mol) | 318.50 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction