missing translation for 'onlineSavingsMsg'
Learn More

ALLO-2, MedChemExpress

Product Code. 16497627
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497627 5 mg 5mg
16484878 10 mg 10mg
16421137 50 mg 50mg
16405666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497627 Supplier Medchem Express Supplier No. HY117407
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ALLO-2 is a potent drug-resistant Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with IC50 of 6 nM.

ALLO-2 is a potent drug-resistant Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with IC50 of 6 nM.

Specifications

Chemical Name or Material ALLO-2
CAS 1357350-60-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H12F3N5O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (336.64 mM; Need ultrasonic)
SMILES FC(F)(OC1=CC=C(C=C1)C2=NC(NC3=CC4=C(C=C3)NN=C4)=NC=C2)F
Molecular Weight (g/mol) 371.32
Formula Weight 371.32
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.