missing translation for 'onlineSavingsMsg'
Learn More

Allitinib tosylate, MedChemExpress

Product Code. 16471847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471847 50 mg 50mg
16428377 5 mg 5mg
16405688 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471847 Supplier Medchem Express Supplier No. HY13427
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Allitinib tosylate (AST-1306 (TsOH)) is an orally active and irreversible EGFR and ErbB2 inhibitor with IC50s of 0.5 and 3 nM, respectively. Allitinib tosylate also inhibits ErbB4 with an IC50 of 0.8 nM. Allitinib tosylate is an anilino-quinazoline compound and has anti-cancer activity

Allitinib tosylate (AST-1306 (TsOH)) is an orally active and irreversible EGFR and ErbB2 inhibitor with IC50s of 0.5 and 3 nM, respectively. Allitinib tosylate also inhibits ErbB4 with an IC50 of 0.8 nM. Allitinib tosylate is an anilino-quinazoline compound and has anti-cancer activity

Specifications

Chemical Name or Material Allitinib tosylate
CAS 1050500-29-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H26ClFN4O5S
Quantity 50 mg
Synonym AST-1306 (TsOH)
Solubility Information DMSO : 50 mg/mL (80.50 mM; Need ultrasonic)
SMILES FC1=CC=CC(COC2=C(Cl)C=C(NC3=NC=NC4=C3C=C(NC(C=C)=O)C=C4)C=C2)=C1.O=S(C5=CC=C(C)C=C5)(O)=O
Molecular Weight (g/mol) 621.08
Formula Weight 621.08
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.