missing translation for 'onlineSavingsMsg'
Learn More

Allitinib tosylate, MedChemExpress

Product Code. 16428377
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16428377

Brand: Medchem Express HY13427

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Allitinib tosylate (AST-1306 (TsOH)) is an orally active and irreversible EGFR and ErbB2 inhibitor with IC50s of 0.5 and 3 nM, respectively. Allitinib tosylate also inhibits ErbB4 with an IC50 of 0.8 nM. Allitinib tosylate is an anilino-quinazoline compound and has anti-cancer activity

Allitinib tosylate (AST-1306 (TsOH)) is an orally active and irreversible EGFR and ErbB2 inhibitor with IC50s of 0.5 and 3 nM, respectively. Allitinib tosylate also inhibits ErbB4 with an IC50 of 0.8 nM. Allitinib tosylate is an anilino-quinazoline compound and has anti-cancer activity

Specifications

Chemical Name or Material Allitinib tosylate
CAS 1050500-29-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H26ClFN4O5S
Quantity 5 mg
Synonym AST-1306 (TsOH)
Solubility Information DMSO : 50 mg/mL (80.50 mM; Need ultrasonic)
SMILES FC1=CC=CC(COC2=C(Cl)C=C(NC3=NC=NC4=C3C=C(NC(C=C)=O)C=C4)C=C2)=C1.O=S(C5=CC=C(C)C=C5)(O)=O
Molecular Weight (g/mol) 621.08
Formula Weight 621.08
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.