missing translation for 'onlineSavingsMsg'
Learn More

Allergin-1 Antibody, Novus Biologicals™

Product Code. 18396291 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396291 100 μL 100µL
18694360 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18396291 Supplier Novus Biologicals Supplier No. NBP276516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Allergin-1 Polyclonal specifically detects Allergin-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Allergin-1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, Probable Mast Cell Antigen 32 Homolog
Gene Symbols MILR1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RKAVLDCEAMKTNEFPSPCLDSKTKVVMKGQNVSMFCSHKNKSLQITYSLFRRKTHLGTQDGKGEPAIFNLSITEAHESGPYKCKAQVTSCSKYSRDFSFTIVDPVTSPVLNIMVIQTETDRHITLHCLSVNGSLP
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 284021.0
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.