missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, MedChemExpress

Product Code. 16439106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439106 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439106 Supplier Medchem Express Supplier No. HYN0543
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Allantoin is a skin conditioning agent that promotes healthy skin, stimulates new and healthy tissue growth.

Allantoin is a skin conditioning agent that promotes healthy skin, stimulates new and healthy tissue growth.

Specifications

Chemical Name or Material Allantoin
CAS 97-59-6
Color White
Purity Grade Notes Research
Molecular Formula C4H6N4O3
Quantity 100 mg
Synonym 5-Ureidohydantoin
Solubility Information DMSO : 50 mg/mL (316.22 mM; Need ultrasonic) ∣H2O : 3.85 mg/mL (24.35 mM; Need ultrasonic)
SMILES O=C(N)NC(C(N1)=O)NC1=O
Molecular Weight (g/mol) 158.12
Formula Weight 158.12
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.