missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, LoGiCal

Product Code. 30458730
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30458730 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30458730

Brand: LoGiCal LGCFOR1627.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allantoin
CAS 97-59-6
Analyte or Component Names Allantoin
Shipping Condition Room Temperature
Molecular Formula C4 H6 N4 O3
Certifications/Compliance ISO 9001
Synonym Allantoin, (RS)-(2,5-dioxoimidazolidin-4-yl)urea, Urea, N-(2,5-dioxo-4-imidazolidinyl)-Allantoin (8CI), Urea, (2,5-dioxo-4-imidazolidinyl)- (9CI), (2,5-Dioxo-4-imidazolidinyl)urea, 5-Ureidohydantoin, Allantion, Allantol, Cordianine, DL-Allantoin, Glyoxyldiureid, Glyoxyldiureide, Glyoxylic diureide, NSC 7606, Psoralon, Sebical, Septalan, (+/-)-Allantoin
InChI Formula InChI=1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)
Solution Type Neat
SMILES NC(=O)NC1NC(=O)NC1=O
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
Industry Type Clinical Testing
Molecular Weight (g/mol) 158.12
Quantity 10 mg
Formula Weight 158.044 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.