Learn More
Brady™ All Weather Permanent Adhesive Green Vinyl Label Tape for M6/M7 Printers
With the ability to last up to 10 years in an outdoor environment, Brady™’s low shrink vinyl with ultra-aggressive adhesive is ideal for a wide range of identification applications.
Brand:  Brady™ 174136
 View more versions of this product
Product Code. 17808864
  • ONLINE EXCLUSIVE PRICE
    £101.00
    £96.14 / Pack of 1
    Save £4.86
    4% Off
Estimated Shipment: 30-11-2026
Add to basket
Description
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty, and harsh environment surfaces, and can be applied in cold temperatures
  • Resists exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and general industrial grime
  • Besides powder coated metal, also adheres to difficult surfaces such as painted cinder block, painted wood, textured plastic, paper-jacketed pipes, and blow-molded cases
  • RoHS compliant to 2011/65/EU (Pass 2015/863)
  • Made of B-595 vinyl material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Green
Continuous
Single-side Printable
Label
Equipment Labeling, Electrical Labeling, Floor Marking, Laboratory Labeling, Lean and 5S Labeling
No
Rectangle
-40°C to +82°C
0.102 mm
50 ft.
2 in.
M6C-2000-595-GN
ASTM D 2979, ASTM D 523
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 ft.
-40°F to +180°F
0.004 in.
50 ft. x 2 in.
15.24 m
50.8 mm