missing translation for 'onlineSavingsMsg'
Learn More

all-trans-Retinyl Palmitate, TRC

Product Code. 30412451
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412451 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412451 Supplier Toronto Research Chemicals Supplier No. TRCR27545025G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vitamin A palmitate (Retinol palmitate)
CAS 79-81-2
Molecular Formula C36 H60 O2
Quantity 25 g
Synonym Retinol palmitate (6CI,7CI), Retinol','palmitate','all-trans- (8CI), 250S/N, 250S/N-B, Aquapalm, Aquasol A, Arovit, Arovit (Roche), Axerophthol palmitate, Dispatabs Tabs, Lutavit A 500 Plus, Myvak, Myvax, Optovit A, Palmitic acid','ester with retinol, Retiblan, Retinyl palmitate, Riken A Palmitate 1000E, Testavol S, VAP 250CK-CWD, VAP 250MS-CWD, Vitamin A palmitate, Vitazyme A, all-trans-Retinol palmitate, all-trans-Retinyl palmitate, all-trans-Vitamin A palmitate, trans-Retinol palmitate, trans-Retinyl palmitate
InChI Formula InChI=1S/C36H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-25-35(37)38-30-28-32(3)23-20-22-31(2)26-27-34-33(4)24-21-29-36(34,5)6/h20,22-23,26-28H,7-19,21,24-25,29-30H2,1-6H3/b23-20+,27-26+,31-22+,32-28+
SMILES CCCCCCCCCCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
Molecular Weight (g/mol) 524.86
Formula Weight 524.4593
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.