missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies All-Trans Retinoic Acid

Product Code. 15855521
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15855521 50 mg 50mg
17110052 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15855521 Supplier STEMCELL Technologies Supplier No. 72262
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Retinoid pathway activator; Activates retinoic acid receptor (RAR)

  • All-Trans Retinoic Acid is a derivative of Vitamin A that functions as a ligand for the retinoic acid receptor (RAR, IC50 = 14 nM)
  • RARs heterodimerize with retinoid X receptors (RXRs) and bind to retinoic acid response elements (RAREs) in DNA and act as transcription factors, altering gene expression
  • (Apfel et al.), Chambon)

Specifications

CAS 302-79-4
Synonym ATRA; NSC 122758; Retinoic acid; Trans retinoic acid; Tretinoin; Vitamin A acid
Quantity 50 mg
Molecular Formula C20H28O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.