missing translation for 'onlineSavingsMsg'
Learn More

All-trans-retinal, MedChemExpress

Product Code. 16449316
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16449316

Brand: Medchem Express HYW004500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress All-trans-retinal is a one of the major vitamin A metabolites in the retina. In physiological conditions, all-trans-RAL is regenerated to the visual chromophore, 11-cis-retinal.

All-trans-retinal is a one of the major vitamin A metabolites in the retina. In physiological conditions, all-trans-RAL is regenerated to the visual chromophore, 11-cis-retinal.

Specifications

Chemical Name or Material All-trans-retinal
CAS 116-31-4
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C20H28O
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (439.46 mM; Need ultrasonic)
SMILES CC(/C=C/C=C(/C=C/C1=C(CCCC(C)1C)C)C)=C\C=O
Molecular Weight (g/mol) 284.44
Formula Weight 284.44
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.