missing translation for 'onlineSavingsMsg'
Learn More

all-trans 4-Keto Retinoic Acid-(9-methyl)-d3, TRC

Product Code. 30389517
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389517 10 mg
30400523 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389517 Supplier Toronto Research Chemicals Supplier No. TRCK20498310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346606-26-5
Molecular Formula C20 2H3 H23 O3
Molecular Weight (g/mol) 317.44
Synonym 4-Oxoretinoic-(9-methyl-d3) acid, 4-Ketoretinoic acid (9-methyl-D3), 4-Oxo-all-trans-retinoic-d3 acid, 4-Oxotretinoin-d3, 4-keto-Retinoic acid-d3, all-trans-4-Oxoretinoic acid-d3, (2E,4E,6E,8E)-3-Methyl-7-(methyl-d3)-9-(2,6,6-trimethyl-3-oxocyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
IUPAC Name (2E,4E,6E,8E)-3-methyl-7-(trideuteriomethyl)-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES [2H]C([2H])([2H])\C(=C/C=C/C(=C/C(=O)O)/C)\C=C\C1=C(C)C(=O)CCC1(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13+/i1D3
Formula Weight 317.207
Chemical Name or Material all-trans-4-Ketoretinoic acid 9-methyl-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.