missing translation for 'onlineSavingsMsg'
Learn More

all-trans-13,14-Dihdyro Retinol (>80%), TRC

Product Code. 30380790
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380790 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380790 Supplier Toronto Research Chemicals Supplier No. TRCD4493505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material all-trans-13,14-Dihdyroretinol
CAS 115797-14-3
Molecular Formula C20 H32 O
Quantity 5 mg
Synonym Retinol, 13,14-dihydro-, 13,14-Dihydroretinol, 13,14-Dihydro-all-trans-retinol, all-trans-13,14-Dihydroretinol
InChI Formula InChI=1S/C20H32O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-12,17,21H,7,10,13-15H2,1-5H3/b9-6+,12-11+,16-8+
SMILES CC(CCO)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
IUPAC Name (4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-4,6,8-trien-1-ol
Molecular Weight (g/mol) 288.47
Formula Weight 288.2453
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.