missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ALKBH5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£189.00 - £274.00
Specifications
| Antigen | ALKBH5 |
|---|---|
| Dilution | Western Blot 1:500-2000, Immunohistochemistry-Paraffin 1:50-200 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041625
|
Novus Biologicals™
NBP3-27856-50ul |
50 μL |
£189.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041670
|
Novus Biologicals™
NBP3-27856-100ul |
100 μL |
£274.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALKBH5 Polyclonal antibody specifically detects ALKBH5 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
| ALKBH5 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Direct Reversal of DNA Damage | |
| PBS, 50% glycerol, 0.5% BSA | |
| 54890 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot 1:500-2000, Immunohistochemistry-Paraffin 1:50-200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ABH5OFOXD, alkB, alkylation repair homolog 5 (E. coli), Alkylated DNA repair protein alkB homolog 5, EC 1.14.11.-, FLJ20308, OFOXD1, oxoglutarate and iron-dependent oxygenase domain containing, probable alpha-ketoglutarate-dependent dioxygenase ABH5 | |
| Synthesized peptide derived from human ALKBH5 (Uniprot #: Q6P6C2) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title