missing translation for 'onlineSavingsMsg'
Learn More

ALKBH5 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18415435 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415435 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415435 Supplier Novus Biologicals Supplier No. NBP278817FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ALKBH5 Polyclonal antibody specifically detects ALKBH5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH5
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias ABH5OFOXD, alkB, alkylation repair homolog 5 (E. coli), Alkylated DNA repair protein alkB homolog 5, EC 1.14.11.-, FLJ20308, OFOXD1, oxoglutarate and iron-dependent oxygenase domain containing, probable alpha-ketoglutarate-dependent dioxygenase ABH5
Host Species Rabbit
Immunogen Partial recombinant protein made to an N-terminal portion of the human ALKB5H protein (amino acids 1-394) [UniProt 54890].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 54890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.