missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALKBH4 Recombinant Rabbit Monoclonal Antibody (24GB6805)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | ALKBH4 |
|---|---|
| Clone | 24GB6805 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275083
![]() |
Invitrogen™
MA556963 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274845
|
Invitrogen™
MA556964 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALKBH4 Recombinant Monoclonal Antibody for Western Blot
Dioxygenase that mediates demethylation of actin monomethylated at 'Lys-84' (K84me1), thereby acting as a regulator of actomyosin-processes (PubMed:23673617). Demethylation of actin K84me1 is required for maintaining actomyosin dynamics supporting normal cleavage furrow ingression during cytokinesis and cell migration (PubMed:23673617). In addition to proteins, also demethylates DNA: specifically demethylates DNA methylated on the 6th position of adenine (N(6)-methyladenosine) DNA, thereby regulating Polycomb silencing. [UniProt]Specifications
| ALKBH4 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| ALKBH4 | |
| 2010004B12Rik; ABH4; Abj4; alkB homolog 4, lysine demethylase; alkB homolog 4, lysine demthylase; alkB, alkylation repair homolog 4; alkB, alkylation repair homolog 4 (E. coli); Alkbh4; Alkylated DNA repair protein alkB homolog 4; alpha-ketoglutarate-dependent dioxygenase alkB homolog 4; DNA N6-methyl adenine demethylase ALKBH4; Lysine-specific demethylase ALKBH4; probable alpha-ketoglutarate-dependent dioxygenase ABH4; RGD1308608 | |
| ALKBH4 | |
| Primary | |
| Affinity chromatography |
| 24GB6805 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NXW9 | |
| 54784 | |
| Recombinant protein of human ALKBH4 (aa 2-302). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title