missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552672
This item is not returnable.
View return policy
Beskrivning
Immunogen sequence: WAAPVKSQAI AQPATTAKSH LHQKPGQTWK NKEHHLSDRE FVFKEPQQVV RRAPEPRVID REGVYEISLS PTGVSRVCLY PGFVDVKEAD WILEQLCQDV PWKQRTGIRE DITYQQPRLT AWYGEL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
ALKBH3 Polyclonal antibody specifically detects ALKBH3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifikationer
| ALKBH3 | |
| Polyclonal | |
| Unconjugated | |
| ALKBH3 | |
| 1700108H04Rik; 1810020C19Rik; Abh3; AlkB homolog 3; alkB homolog 3, alpha-ketoglutaratedependent dioxygenase; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; alkB, alkylation repair homolog 3 (E. coli); Alkbh3; Alkylated DNA repair protein alkB homolog 3; alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC 1; DEPC1; DEPC-1; FLJ43614; hABH3; m MGC118790; mABH3; MGC118792; MGC118793; MGC134125; PCA1; Prostate cancer antigen 1; prostate cancer antigen-1; RP23-375N21.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221120 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96Q83 | |
| ALKBH3 | |
| Recombinant protein corresponding to Human ALKBH3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering