missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALKBH3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576827
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Dioxygenase that repairs alkylated DNA containing 1- methyladenine (1meA) and 3-methylcytosine (3meC) by oxidative demethylation. Has a strong preference for single-stranded DNA. Able to process alkylated 3mC within double-stranded regions via its interaction with ASCC3, which promotes DNA unwinding to generate single-stranded substrate needed for ALKHB3. May also act on RNA. Requires molecular oxygen, alpha-ketoglutarate and iron.
Specifications
| ALKBH3 | |
| Polyclonal | |
| Unconjugated | |
| Alkbh3 | |
| 1700108H04Rik; 1810020C19Rik; Abh3; AlkB homolog 3; alkB homolog 3, alpha-ketoglutaratedependent dioxygenase; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; alkB, alkylation repair homolog 3 (E. coli); Alkbh3; Alkylated DNA repair protein alkB homolog 3; alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC 1; DEPC1; DEPC-1; FLJ43614; hABH3; m MGC118790; mABH3; MGC118792; MGC118793; MGC134125; PCA1; Prostate cancer antigen 1; prostate cancer antigen-1; RP23-375N21.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221120, 362169, 69113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XIC8, Q8K1E6, Q96Q83 | |
| Alkbh3 | |
| Recombinant full length Human ALKBH3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction