missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38187-20ul
This item is not returnable.
View return policy
Description
ALKBH3 Polyclonal antibody specifically detects ALKBH3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| ALKBH3 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:4000, ELISA | |
| ABH3MGC118793, alkB, alkylation repair homolog 3 (E. coli), Alkylated DNA repair protein alkB homolog 3, DEPC1alpha-ketoglutarate-dependent dioxygenase alkB homolog 3, DEPC-1MGC118792, EC 1.14.11, EC 1.14.11.-, FLJ43614, MGC118790, PCA1, Prostate cancer antigen 1, prostate cancer antigen-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-286 of human ALKBH3 (NP_631917.1).,, Sequence:, MEEKRRRARVQGAWAAPVKSQAIAQPATTAKSHLHQKPGQTWKNKEHHLSDREFVFKEPQQVVRRAPEPRVIDREGVYEISLSPTGVSRVCLYPGFVDVKEADWILEQLCQDVPWKQRTGIREDITYQQPRLTAWYGELPYTYSRITMEPNPHWHPVLRTLKNRIEENTGHTFNSLLCNLYRNEKDSVDWHSDDEPSLGRCPIIASLSFGATRTFEMRKKPPPEENGDYTYVERVKIPLDHGTLLIMEGATQADWQHRVPKEYHSREPRVNLTFRTVYPDPRGAPW | |
| 20 μL | |
| Proteases & Other Enzymes | |
| 221120 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction