missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38187-100ul
This item is not returnable.
View return policy
Description
ALKBH3 Polyclonal antibody specifically detects ALKBH3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| ALKBH3 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:4000, ELISA | |
| ABH3MGC118793, alkB, alkylation repair homolog 3 (E. coli), Alkylated DNA repair protein alkB homolog 3, DEPC1alpha-ketoglutarate-dependent dioxygenase alkB homolog 3, DEPC-1MGC118792, EC 1.14.11, EC 1.14.11.-, FLJ43614, MGC118790, PCA1, Prostate cancer antigen 1, prostate cancer antigen-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-286 of human ALKBH3 (NP_631917.1).,, Sequence:, MEEKRRRARVQGAWAAPVKSQAIAQPATTAKSHLHQKPGQTWKNKEHHLSDREFVFKEPQQVVRRAPEPRVIDREGVYEISLSPTGVSRVCLYPGFVDVKEADWILEQLCQDVPWKQRTGIREDITYQQPRLTAWYGELPYTYSRITMEPNPHWHPVLRTLKNRIEENTGHTFNSLLCNLYRNEKDSVDWHSDDEPSLGRCPIIASLSFGATRTFEMRKKPPPEENGDYTYVERVKIPLDHGTLLIMEGATQADWQHRVPKEYHSREPRVNLTFRTVYPDPRGAPW | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 221120 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur