missing translation for 'onlineSavingsMsg'
Learn More

ALKBH3 Antibody, Novus Biologicals™

Product Code. 18393957 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393957 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393957 Supplier Novus Biologicals Supplier No. NBP312996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ALKBH3 Polyclonal antibody specifically detects ALKBH3 in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias ABH3MGC118793, alkB, alkylation repair homolog 3 (E. coli), Alkylated DNA repair protein alkB homolog 3, DEPC1alpha-ketoglutarate-dependent dioxygenase alkB homolog 3, DEPC-1MGC118792, EC 1.14.11, EC 1.14.11.-, FLJ43614, MGC118790, PCA1, Prostate cancer antigen 1, prostate cancer antigen-1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALKBH3. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 221120
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.